Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GARNL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554630
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554630 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554630 Supplier Invitrogen™ Supplier No. PA554630
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDPENVGTPT SLGSSICEEE EEDNLSPNTF GYKLECKGEA RAYRRHFLGK DHLNFYCTGS SLGNLILSVK CEEAEGIEYL RVILRSKLKT VHERIPLAGL SKLPSVPQIA KAFCDDAVGL RFNPVLYPKA SQMIVSYDEH Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GARNL4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAP1GAP2
Gene Accession No. Q684P5
Gene Alias GARNL4; GTPase activating RANGAP domain-like 4; GTPase activating Rap/RanGAP domain-like 4; GTPase-activating Rap/Ran-GAP domain-like protein 4; KIAA1039; RAP1 GTPase activating protein 2; rap1 GTPase-activating protein 2; RAP1GA2; RAP1GA3; Rap1GAP2
Gene Symbols RAP1GAP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GARNL4. Recombinant protein control fragment (Product #RP-93264).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23108, 303298
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.