Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GARP Monoclonal Antibody (G14D9), Super Bright™ 702, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$417.00
Specifications
Antigen | GARP |
---|---|
Clone | G14D9 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
67-988-242
![]() |
Invitrogen™
67988242 |
100 Tests |
Each of 1 for $417.00
|
|
|||||
Description
Rat Monoclonal Antibody
GARP, also known as leucine rich repeat containing 32, is a type I membrane protein with twenty-two leucine-rich repeats and a short intracytoplasmic region. GARP was identified based on the analysis of gene expression profiling of Treg and Th cells following TCR stimulation. Since GARP is expressed in CD4+, CD25+, FoxP3+ cells (Tregs), it may be a candidate for cell surface marker for Treg cells.Specifications
GARP | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
2615 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
G14D9 | |
Flow Cytometry | |
Super Bright 702 | |
Rat | |
Human | |
Q14392 | |
LRRC32 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
LRRC32 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title