Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GARP Monoclonal Antibody (YGIC86), PE, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Supplier:  Invitrogen™ 12989182

Encompass_Preferred

 View more versions of this product

Catalog No. 12-989-182


Get 15% Off Invitrogen Antibodies and Reagents
Use promo code LABELING15 at checkout
View Available Promotions
Only null left
Add to Cart

Description

Description

Description: The YGIC86 monoclonal antibody reacts with mouse Glycoprotein A Repetitions Predomit (GARP, also known as Lrrc32 or Garpin). GARP is an approximately 80 kDa glycoprotein that is expressed on the cell surface. Using northern blot, RT-PCR or microarray analyses, the expression of GARP has been reported in placenta, lung, kidney, heart, ovary, liver, skeletal muscle, and pancreas. Protein expression has been observed on megakaryocytes, platelets and activated regulatory T (Treg) cells. The expression of GARP on Treg cells has been reported to be necessary for the suppressive function of Treg cells, possibly related to its role as a cell surface receptor for LAP/TGF beta. When looking for expression of GARP on platelets, it is recommended to use the Foxp3 Staining Buffer Set (cat. 00-5523) to fix the platelets before staining with this YGIC86 monoclonal antibody. Fixation/permeabilization is not necessary when staining Treg cells. YGIC86 does not cross-react to rat GARP. Applications Reported: This YGIC86 antibody has been reported for use in flow cytometric analysis. Applications Tested: This YGIC86 antibody has been tested by flow cytometric analysis of stimulated mouse spleen cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.

GARP, also known as leucine rich repeat containing 32, is a type I membrane protein with twenty-two leucine-rich repeats and a short intracytoplasmic region. GARP was identified based on the analysis of gene expression profiling of Treg and Th cells following TCR stimulation. Since GARP is expressed in CD4+, CD25+, FoxP3+ cells (Tregs), it may be a candidate for cell surface marker for Treg cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GARP
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
G3XA59
LRRC32
Affinity chromatography
RUO
434215
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
YGIC86
PE
LRRC32
AI426318; D11S833E; D11S833Eh; D7H11S833E; EG434215; Garp; garpin; Glycoprotein A repetitions predominant; leucine rich repeat containing 32; leucine-rich repeat-containing protein 32; Lrrc32; Transforming growth factor beta activator LRRC32
Rat
100 μg
Primary
Mouse
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only.