Learn More
GARP Monoclonal Antibody (YGIC86), Super Bright™ 436, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$476.00
Specifications
Antigen | GARP |
---|---|
Clone | YGIC86 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
62-989-182
![]() |
Invitrogen™
62989182 |
100 μg |
Each of 1 for $476.00
|
|
|||||
Description
The YGIC86 monoclonal antibody reacts with mouse Glycoprotein A Repetitions Predomit (GARP, also known as Lrrc32 or Garpin). GARP is an approximately 80 kDa glycoprotein that is expressed on the cell surface. Using northern blot, RT-PCR or microarray anal
GARP, also known as leucine rich repeat containing 32, is a type I membrane protein with twenty-two leucine-rich repeats and a short intracytoplasmic region. GARP was identified based on the analysis of gene expression profiling of Treg and Th cells following TCR stimulation. Since GARP is expressed in CD4+, CD25+, FoxP3+ cells (Tregs), it may be a candidate for cell surface marker for Treg cells.Specifications
GARP | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
434215 | |
Primary | |
Antibody |
YGIC86 | |
Flow Cytometry | |
Super Bright 436 | |
Rat | |
Mouse | |
IgG2a κ | |
4°C, store in dark, DO NOT FREEZE! |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.