Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GARP Mouse anti-Mouse, Bovine, Unconjugated, Clone: 4B1, MilliporeSigma™

Catalog No. MABN242925 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2429-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN2429-25 Supplier MilliporeSigma Supplier No. MABN242925UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-GARP, clone 4B1, Cat. No. MABN2429, is a highly specific mouse monoclonal antibody, that targets CNGB1 and has been tested in Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GARP
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 4B1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Cyclic nucleotide-gated cation channel beta-1;Cyclic nucleotide-gated cation channel 4;CNG channel 4;CNG-4;CNG4;Cyclic nucleotide-gated cation channel gamma;Cyclic nucleotide-gated cation channel modulatory subunit;Cyclic nucleotide-gated channel beta-1;CNG channel beta-1;Glutamic acid-rich protein;GARP
Gene Symbols CNGBI;CNCG2;CNCG3L;CNCG4;RCNC2
Host Species Mouse
Immunogen GST-tagged first 16 amino acids from the N-terminal region of human Cyclic nucleotide-gated cation channel beta-1.
Molecular Weight of Antigen 139.68 kDa calculated.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone 4B1 detects Glutamic acid-rich protein (GARP) in mouse and bovine retina. It targets an epitope with in the first 16 amino acids from the N-terminal region.
Target Species Mouse, Bovine
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.