Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GARS Antibody (4E10), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242679F
Description
GARS Monoclonal specifically detects GARS in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
GARS | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
GARS | |
Recombinant human GARS (43-289aa) purified from E. coli | |
100 μL | |
Core ESC Like Genes, Stem Cell Markers | |
2617 | |
Store at 4C in the dark. | |
IgG1 κ |
ELISA, Immunocytochemistry, Immunofluorescence | |
40000000000 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
AP-4-A synthetase, Charcot-Marie-Tooth neuropathy 2D, Charcot-Marie-Tooth neuropathy, neuronal type, D, Diadenosine tetraphosphate synthetase, DSMAV, glycine tRNA ligase, Glycine--tRNA ligase, glycyl-tRNA synthetase, GlyRSEC 6.1.1.14, HMN5, SMAD1CMT2D | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction