Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GARS Antibody (OTI4G4), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270794H
Description
GARS Monoclonal specifically detects GARS in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
GARS | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
GARS | |
Full length recombinant protein of human GARS (NP_002038) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunohistochemistry | |
OTI4G4 | |
Immunohistochemistry | |
AP-4-A synthetase, Charcot-Marie-Tooth neuropathy 2D, Charcot-Marie-Tooth neuropathy, neuronal type, D, Diadenosine tetraphosphate synthetase, DSMAV, glycine tRNA ligase, Glycine--tRNA ligase, glycyl-tRNA synthetase, GlyRSEC 6.1.1.14, HMN5, SMAD1CMT2D | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
2617.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction