Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GARS Monoclonal Antibody (OTI4H8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA805076

Description
This gene encodes glycyl-tRNA synthetase, one of the aminoacyl-tRNA synthetases that charge tRNAs with their cognate amino acids. The encoded enzyme is an (alpha)2 dimer which belongs to the class II family of tRNA synthetases. It has been shown to be a target of autoantibodies in the human autoimmune diseases, polymyositis or dermatomyositis.Spécifications
GARS | |
Monoclonal | |
Unconjugated | |
GARS | |
CMT2D, DSMAV, GlyRS, HMN5, SMAD1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
2617 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4H8 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P41250 | |
GARS | |
Full length human recombit protein of human GARS produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
missing translation for 'provideContentCorrection'
missing translation for 'type07FormHeader'
missing translation for 'productTitle'
missing translation for 'spotOppurtunityImprovement'missing translation for 'shareContentCorrection'