Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BINDER™ Gas-tight Option Model
Shop All BINDER Inc. Products

Click to view available options
For Use With (Equipment):
Units M 115
Units M 260
Units M 56
Description
- Gas-tight option model
- For use with various BINDER™ Unit M lines
Specifications
Specifications
Description | Gas-tight Option Model |
For Use With (Equipment) | Units M 56 |
Material | Steel |
Weight (English) | 2.20 lb. |
Weight (Metric) | 1 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction