Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gas1 Antibody (11C47), DyLight 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP222032UV
Description
Gas1 Monoclonal specifically detects Gas1 in Human samples. It is validated for Western Blot, ELISA.Specifications
Gas1 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GAS1 | |
Human recombinant Gas1 | |
0.1 mL | |
2619 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, ELISA | |
11C47 | |
Western Blot, ELISA | |
GAS-1, growth arrest-specific 1, Growth arrest-specific gene-1, growth arrest-specific protein 1 | |
Rat | |
Protein G purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction