Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GAS1 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547360
Description
In sandwich ELISAs, less than 0.1% cross-reactivity with recombinant human Gas1 and recombinant mouse Gas6 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Growth arrest-specific 1 plays a role in growth suppression. GAS1 blocks entry to S phase and prevents cycling of normal and transformed cells. Gas1 is a putative tumor suppressor gene.
Specifications
GAS1 | |
Polyclonal | |
Unconjugated | |
GAS1 | |
AW554192; GAS1; Gas-1; growth arrest specific 1; growth arrest-specific 1; Growth arrest-specific gene-1; growth arrest-specific protein 1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
14451 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
ELISA, Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q01721 | |
GAS1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Gas1 Leu39-Ser315. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction