Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GAS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564296
Description
Immunogen sequence: TVIEDMLAMP KAALLNDCVC DGLERPICES VKENMARLCF GAELGNGPGS SGSDGGLDDY YDEDYDDEQR TGGAGGEQPL DDDDG Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
Growth arrest-specific 1 plays a role in growth suppression. GAS1 blocks entry to S phase and prevents cycling of normal and transformed cells. Gas1 is a putative tumor suppressor gene.
Specifications
GAS1 | |
Polyclonal | |
Unconjugated | |
GAS1 | |
AW554192; GAS1; Gas-1; growth arrest specific 1; growth arrest-specific 1; Growth arrest-specific gene-1; growth arrest-specific protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2619 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P54826 | |
GAS1 | |
Recombinant protein corresponding to Human GAS1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction