Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GAS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565166
Description
Immunogen sequence: TVIEDMLAMPK AALLNDCVCD GLERPICESV KENMARLCFG AELGNGPGSS GSDGGLDDYY DEDYDDEQRT GGAGGEQPLD DDDG Highest antigen sequence identity to the following orthologs - mouse 89%, rat 89%.
Growth arrest-specific 1 plays a role in growth suppression. GAS1 blocks entry to S phase and prevents cycling of normal and transformed cells. Gas1 is a putative tumor suppressor gene.
Specifications
GAS1 | |
Polyclonal | |
Unconjugated | |
GAS1 | |
AW554192; GAS1; Gas-1; growth arrest specific 1; growth arrest-specific 1; Growth arrest-specific gene-1; growth arrest-specific protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2619 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P54826 | |
GAS1 | |
Recombinant Human GAS1. Recombinant protein control fragment (Product #RP-104295). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction