Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAS2 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298799F
Description
GAS2 Polyclonal specifically detects GAS2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
GAS2 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
GAS-2, growth arrest-specific 2, growth arrest-specific protein 2, MGC32610 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human GAS2. | |
0.1 mL | |
Apoptosis | |
2620 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
FITC | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction