Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAS2 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298799JF646
Description
GAS2 Polyclonal specifically detects GAS2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| GAS2 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| GAS-2, growth arrest-specific 2, growth arrest-specific protein 2, MGC32610 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human GAS2. | |
| 0.1 mL | |
| Apoptosis | |
| 2620 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction