Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAS2 Antibody (SR1577), Novus Biologicals™
SDP

Catalog No. NB258445 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258445 50 μg
NB258433 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB258445 Supplier Novus Biologicals Supplier No. NBP32261550UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GAS2 Monoclonal antibody specifically detects GAS2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Serum

Specifications

Antigen GAS2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone SR1577
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry, Serum 1:50-1:100
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias GAS-2, growth arrest-specific 2, growth arrest-specific protein 2, MGC32610
Host Species Rabbit
Immunogen A synthesized peptide derived from human GAS2 (Uniprot #: O43903)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 2620
Target Species Human, Mouse
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.