Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAS2L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582704
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582704 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582704 Supplier Invitrogen™ Supplier No. PA582704
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLSRVSSPSP ELGTTPASIF RTPLQLDPQQ EQQLFRRLEE EFLANARALE AVASVTPTGP APDPARAPDP PAPDSAYCSS SSSSSSLSVL GGKCG Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 32%.

The Gas2 protein is thought to play a role in apoptosis by acting as a cell death substrate for caspases. Gas2, a component of the microfilament system, is cleaved by a caspase (caspase-3 or caspase-7) at Asparagine 278 during apoptosis. The cleaved form of Gas2 dramatically induces the re-arrangement of the Actin cytoskeleton and causes potent changes in the shape of the affected cells. Gas2 is a cytoskeleton and peripheral membrane protein that co-localizes with Actin fibers at the cell border and along the stress fibers in growth-arrested fibroblasts. Gas2 is mainly membrane-associated, but when hyperphosphorylated it will accumulate at membrane ruffles, indicating Gas2 involvement in that process. Gar22 (Gas2-related protein on chromosome 22), also designated growth arrest-specific protein 2-like 1 (Gas2-like1), belongs to the Gas2 family and contains one CH (calponinhomo-logy) domain. It is thought to be involved in the cross-linking of microtubules and microfilaments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAS2L1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GAS2L1
Gene Accession No. Q99501
Gene Alias 4930500E24Rik; D0Jmb1; Gar22; GAS2L1; GAS2-like protein 1; GAS2-related protein; GAS2-related protein on chromosome 22; growth arrest specific 2 like 1; growth arrest-specific 2 like 1; growth arrest-specific protein 2-like 1; TU-71.1
Gene Symbols GAS2L1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GAS2L1. Recombinant protein control fragment (Product #RP-108158).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10634
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.