Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAS2L1, Rabbit anti-Human, Polyclonal, Millipore Sigma™
Rabbit Polyclonal Antibody
$192.34 - $498.79
Specifications
Antigen | GAS2L1 |
---|---|
Dilution | Western Blotting Analysis: A representative lot detected GAS2L1 in Western Blotting applications (Au, F.K., et. al. (2017). Dev Cell. 40(1):81-94).Immunoprecipitation Analysis: 1μg from a representative lot detected GAS2L1 in U2OS cell lysate (Courtesy of Dr Robert Z. Qi and Dr. Franco K.C. Au at Hong Kong University of Science and Technology).Immunofluorescence Analysis: A representative lot detected GAS2L1 in Immunofluorescence applications (Au, F.K., et. al. (2017). Dev Cell. 40(1):81-94).Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected GAS2L1 in lysate from HeLa cells, but not in lysate from GAS2L1 knockout cells (Courtesy of Dr Robert Z. Qi and Dr. Franco K.C. Au at Hong Kong University of Science and Technology). |
Applications | Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABN226125UG
|
MilliporeSigma
ABN226125UG |
25 μg |
Each for $192.34
|
|
|||||
ABN2261100
|
MilliporeSigma
ABN2261100UG |
100 μg |
Each for $498.79
|
|
|||||
Description
Anti-GAS2L1, Cat. No. ABN2261, is a highly specific rabbit polyclonal antibody that targets GAS2-like protein 1 (Gas2L1) and has been tested for use in Immunofluorescence, Immunoprecipitation, and Western Blotting.
GAS2-like protein 1 (UniProt: Q99501-1; also known as GAS2-related protein on chromosome 22, Growth arrest-specific protein 2-like 1, GAS2L1) is encoded by the GAS2L1 (also known as GAR22) gene (Gene ID: 10634) in human. GAS2L1 belongs to the growth-arrest-specific 2 (GAS2) family, whose members mediate the crosstalk between filamentous actin (F-actin) and microtubules. Three members of this family, known as GAS2L1, GAS2L2, and GAS2L3, are shown to be differentially involved in mediating the crosstalk between F-actin and microtubules. GAS2L1 is expressed in testis and brain, GAS2L2 is exclusively expressed in skeletal muscle, and GAS2L3 is found in many different cell types. Although they all localize to actin and microtubules, only the exogenous expression of GAD2L1 and GAS2L2 affect microtubule stability, dynamics, and guidance along actin stress fibers. GAS2L1 is reported to associate with the proximal end of mature centrioles and participates in centriole dynamics and centrosome disjunction. Loss of GAS2L1 can inhibit centrosome disjunction in the G2 phase. On the other hand, overexpression of GAS2L1 can induce premature centrosome separation, which also requires its association with actin, microtubules, and the microtubule end-binding proteins. (Ref.: Stroud MJ et al. (2014). J. Cell Sci. 127(12); 2672-2682; AU, FKC et al. (2017). Dev. Cell 40(1); 81-94).Specifications
GAS2L1 | |
Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Neuroscience | |
Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide and 10% glycerol. | |
NP_689422 | |
GST-tagged recombinant fragment corresponding to 188 amino acids from the C-terminal region of human GAS2-Like protein 1. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
∽78 kDa observed; 72.72 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Western Blotting Analysis: A representative lot detected GAS2L1 in Western Blotting applications (Au, F.K., et. al. (2017). Dev Cell. 40(1):81-94).Immunoprecipitation Analysis: 1μg from a representative lot detected GAS2L1 in U2OS cell lysate (Courtesy of Dr Robert Z. Qi and Dr. Franco K.C. Au at Hong Kong University of Science and Technology).Immunofluorescence Analysis: A representative lot detected GAS2L1 in Immunofluorescence applications (Au, F.K., et. al. (2017). Dev Cell. 40(1):81-94).Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected GAS2L1 in lysate from HeLa cells, but not in lysate from GAS2L1 knockout cells (Courtesy of Dr Robert Z. Qi and Dr. Franco K.C. Au at Hong Kong University of Science and Technology). | |
Polyclonal | |
Purified | |
RUO | |
Human | |
GAS2-like protein 1;GAS2-related protein on chromosome 22;Growth arrest-specific protein 2-like 1 | |
GAS2L1;GAR22 | |
IgG | |
Affinity Purified | |
This rabbit polyclonal antibody GAS2-like protein 1 (GAS2L1) in human cells. It targets an epitope with in 188 amino acids from the C-terminal half. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title