Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAS2L1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | GAS2L1 |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
Description
GAS2L1 Polyclonal specifically detects GAS2L1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GAS2L1 | |
| Unconjugated | |
| RUO | |
| Q99501 | |
| 10634 | |
| Synthetic peptides corresponding to GAS2L1(growth arrest-specific 2 like 1) The peptide sequence was selected from the middle region of human GAS2L1. Peptide sequence ARSQSREEQAVLLVRRDRDGQHSWVPRGRGSGGSGRSTPQTPRARSPAAP. | |
| Primary |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication | |
| GAR22Growth arrest-specific protein 2-like 1, GAS2-like protein 1, GAS2-related protein on chromosome 22, growth arrest-specific 2 like 1, MGC17243 | |
| GAS2L1 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title