Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAS2L3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561776
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561776 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561776 Supplier Invitrogen™ Supplier No. PA561776
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LQFATLEQKI LAFQKGVSNE SVPDSPARTP QPPEMNPLSA VNMFQKQNSK PSVPVSIPKS KEKQGRPPGA LVPASSLKGG NLGSMSVRSK LPNSPAASSH PKLKS Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.

Cytoskeletal linker protein. May promote and stabilize the formation of the actin and microtubule network.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAS2L3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GAS2L3
Gene Accession No. Q86XJ1
Gene Alias 8430435B07Rik; fc11d07; G2L3; GAS2L3; gas2l3b; GAS2-like protein 3; Gm240; growth arrest specific 2 like 3; growth arrest-specific 2 like 3; growth arrest-specific protein 2-like 3; si:ch211-203h15.1; wu:fc11d07
Gene Symbols GAS2L3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GAS2L3. Recombinant protein control fragment (Product #RP-97771).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283431
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.