Learn More
GAS2L3 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304153A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Cytoskeletal linker protein. May promote and stabilize the formation of the actin and microtubule network.Specifications
GAS2L3 | |
Polyclonal | |
Unconjugated | |
GAS2L3 | |
G2L3, growth arrest-specific 2 like 3, growth arrest-specific protein 2-like 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
283431 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q86XJ1 | |
GAS2L3 | |
Between 644 and 694 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.