Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAS41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534867
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534867 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534867 Supplier Invitrogen™ Supplier No. PA534867
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (94%), Xenopus laevis (99%), Chicken (99%), Sheep (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GAS41 has high sequence homology to human MLLT1, and yeast and human MLLT3 proteins. Both MLLT1 and MLLT3 proteins belong to a class of transcription factors, indicating that the encoded protein might also represent a transcription factor. This protein is thought to be required for RNA transcription. This gene has been shown to be amplified in tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAS41
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene YEATS4
Gene Accession No. O95619
Gene Alias 4930573H17Rik; B230215M10Rik; GAS41; Glioma-amplified sequence 41; glioma-amplified sequence-41; nuBI1; NuBI-1; NuMA binding protein 1; nuMA-binding protein 1; RGD1305741; YAF9; YEATS domain containing 4; YEATS domain-containing protein 4; Yeats4
Gene Symbols YEATS4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GAS41. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8089
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.