Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAS41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599230
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599230 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599230 Supplier Thermo Scientific Supplier No. PA599230
Add to Cart
Add to Cart

GAS41 Polyclonal Antibody

GAS41 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen GAS41
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene YEATS4
Gene Accession No. O95619, Q9CR11
Gene Alias 4930573H17Rik; B230215M10Rik; GAS41; Glioma-amplified sequence 41; glioma-amplified sequence-41; nuBI1; NuBI-1; NuMA binding protein 1; nuMA-binding protein 1; RGD1305741; YAF9; YEATS domain containing 4; YEATS domain-containing protein 4; Yeats4
Gene Symbols YEATS4
Host Species Rabbit
Immunogen A synthesized peptide derived from human YEATS4(Accession O95619), corresponding to amino acid residues V63-T113.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64050, 8089
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.