Learn More
GAS41 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA124423
Description
PA1-24423 detects Glioma-Amplified Sequence-41 (YEATS4) in Human samples. PA1-24423 has been successfully used in Western Blot procedures. PA1-24423 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human YEATS4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
YEATS4 is found in the nucleoli. It has high sequence homology to human MLLT1, and yeast and human MLLT3 proteins. Both MLLT1 and MLLT3 proteins belong to a class of transcription factors, indicating that the encoded protein might also represent a transcription factor. This protein is thought to be required for RNA transcription. This gene has been shown to be amplified in tumors.
Specifications
GAS41 | |
Polyclonal | |
Unconjugated | |
O95619 | |
YEATS4 | |
Synthetic peptide region of human YEATS4. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
YEATS4 | |
4930573H17Rik, B230215M10Rik, GAS41, NUBI-1, YAF9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8089 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.