Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gas6 Antibody (RM0084-6J2) - Azide and BSA Free, Novus Biologicals™
SDP

Rat Monoclonal Antibody

$206.50 - $560.50

Specifications

Antigen Gas6
Clone RM0084-6J2
Concentration LYOPH
Dilution Western Blot 1:500-1:1000, ELISA
Applications ELISA, Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018424
SDP
View Documents
Novus Biologicals
NBP1217240.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP121724
SDP
View Documents
Novus Biologicals
NBP121724
0.1 mg
Each for $560.50
Only null left
Add to Cart
 
Description

Description

Gas6 Monoclonal specifically detects Gas6 in Mouse samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Gas6
LYOPH
ELISA, Western Blot
Unconjugated
Rat
Mouse
2621
Mouse recombinant Gas6
Primary
RM0084-6J2
Western Blot 1:500-1:1000, ELISA
Monoclonal
Purified
RUO
AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6
GAS6
IgG2
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.