Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gas6 Antibody (RM0084-6J2), DyLight 650, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP121724C
Description
Gas6 Monoclonal specifically detects Gas6 in Mouse samples. It is validated for Western Blot, ELISA.Specifications
Gas6 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GAS6 | |
Mouse recombinant Gas6 | |
0.1 mL | |
2621 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, ELISA | |
RM0084-6J2 | |
Western Blot, ELISA | |
AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6 | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction