Learn More
Invitrogen™ GAS6 Polyclonal Antibody

Description
Positive test controls include: HeLa, NCI-H460, HT-1080, SW620, DU145. The target is usually found in the following locations: Secreted.
Specifications
Specifications
| Antigen | GAS6 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 3.69 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | GAS6 |
| Gene Accession No. | Q14393, Q61592 |
| Gene Alias | AXL receptor tyrosine kinase ligand; AXL stimulatory factor; AXLLG; AXSF; GAS 6; GAS6; GAS-6; GPF; growth arrest specific 6; growth arrest-specific 6; growth arrest-specific protein 6; growth-potentiating factor |
| Gene Symbols | GAS6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.