Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAS6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552539
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552539 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552539 Supplier Invitrogen™ Supplier No. PA552539
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STWEVEVVAH IRPAADTGVL FALWAPDLRA VPLSVALVDY HSTKKLKKQL VVLAVEHTAL ALMEIKVCDG QEHVVTVSLR DGEATLEVDG TRGQSEVSAA QLQERLAVLE RHLRSPVLTF AGGLPDVPVT S Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 79%.

This gene product is a gamma-carboxyglutamic acid (Gla)-containing protein thought to be involved in the stimulation of cell proliferation, and may play a role in thrombosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAS6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GAS6
Gene Accession No. Q14393
Gene Alias AXL receptor tyrosine kinase ligand; AXL stimulatory factor; AXLLG; AXSF; GAS 6; GAS6; GAS-6; GPF; growth arrest specific 6; growth arrest-specific 6; growth arrest-specific protein 6; growth-potentiating factor
Gene Symbols GAS6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GAS6. Recombinant protein control fragment (Product #RP-102272).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2621
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.