Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GAS6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572882
Description
GAS6 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene product is a gamma-carboxyglutamic acid (Gla)-containing protein thought to be involved in the stimulation of cell proliferation, and may play a role in thrombosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
GAS6 | |
Polyclonal | |
Unconjugated | |
GAS6 | |
AXL receptor tyrosine kinase ligand; AXL stimulatory factor; AXLLG; AXSF; GAS 6; GAS6; GAS-6; GPF; growth arrest specific 6; growth arrest-specific 6; growth arrest-specific protein 6; growth-potentiating factor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14456, 2621, 58935 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q14393, Q61592, Q63772 | |
GAS6 | |
GAS6 antibody for a 19 amino acid peptide from near the amino terminus of human GAS6. The immunogen is within amino acids 60 - 110 of GAS6. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction