Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAS6 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA528227

Catalog No. PIPA528227


Description

Description

PA5-28227 targets GAS6 in IF and WB applications and shows reactivity with Human samples. The PA5-28227 immunogen is recombit fragment corresponding to a region within amino acids 283 and 621 of Human GAS6. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene product is a gamma-carboxyglutamic acid (Gla)-containing protein thought to be involved in the stimulation of cell proliferation, and may play a role in thrombosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GAS6
Polyclonal
Unconjugated
Q14393
GAS6
Recombit fragment corresponding to a region within amino acids 283 and 621 of Human GAS6
100 μL
Primary
Human
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Western Blot
1.02 mg/mL
GAS6
AXLLG, AXSF, AXL receptor tyrosine kinase ligand, AXL stimulatory factor, growth arrest-specific protein 6
Rabbit
Antigen affinity chromatography
RUO
2621
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.