Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAS7 Antibody (OTI2F6), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271376MFV450
Description
GAS7 Monoclonal antibody specifically detects GAS7 in Human, Mouse, Rat, Canine samples. It is validated for Flow CytometrySpecifications
GAS7 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine | |
Purified |
Flow Cytometry | |
OTI2F6 | |
Flow Cytometry | |
GAS-7, growth arrest-specific 7, KIAA0394growth arrest-specific protein 7, MGC1348, MLL/GAS7, MLL/GAS7 fusion protein | |
Full length human recombinant protein of human GAS7 (NP_003635) produced in HEK293T cell. | |
0.1 mL | |
Cell Cycle and Replication | |
8522 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction