Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAS7 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297951PE
Description
GAS7 Polyclonal antibody specifically detects GAS7 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
GAS7 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
GAS-7, growth arrest-specific 7, KIAA0394growth arrest-specific protein 7, MGC1348, MLL/GAS7, MLL/GAS7 fusion protein | |
Produced in rabbits immunized with E. coli-derived Human GAS7 fragment. | |
0.1 mL | |
Cell Cycle and Replication | |
8522 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction