Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAS7 Antibody

Chicken Polyclonal Antibody
Supplier: Novus Biologicals NB10075411
Description
GAS7 Polyclonal specifically detects GAS7 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
GAS7 | |
Polyclonal | |
Western Blot 1:500 | |
O60861 | |
GAS7 | |
The immunogen for this antibody was a sequence from 1a.a. to 412a.a. from Growth arrest specific protein 7 isoform c (a 476a.a. long protein) | |
0.05 mg | |
Cell Cycle and Replication | |
8522 | |
Human, Mouse, Rat |
Western Blot | |
Unconjugated | |
PBS (pH 7.2) with 0.02% Sodium Azide | |
GAS-7, growth arrest-specific 7, KIAA0394growth arrest-specific protein 7, MGC1348, MLL/GAS7, MLL/GAS7 fusion protein | |
Chicken | |
Affinity Purified | |
RUO | |
Primary | |
Growth arrest specific protein 7 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction