Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAS7 Monoclonal Antibody (OTI9H6), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA501873

Encompass_Preferred

 View more versions of this product

Catalog No. 50-167-2980


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

Growth arrest-specific 7 is expressed primarily in terminally differentiated brain cells and predomitly in mature cerebellar Purkinje neurons. GAS7 plays a putative role in neuronal development. Several transcript variants encoding proteins which vary in the N-terminus have been described.
Specifications

Specifications

GAS7
Monoclonal
0.95 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O60861
GAS7
Full length human recombit protein of human GAS7 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2a
Flow Cytometry, Western Blot
OTI9H6
Unconjugated
GAS7
growth arrest specific 7
Mouse
Affinity Chromatography
RUO
8522
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.