Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAS7 Monoclonal Antibody (OTI9H6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501873S


Description
GAS7 Monoclonal antibody specifically detects GAS7 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
GAS7 | |
Monoclonal | |
0.95 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O60861 | |
Mouse | |
Affinity chromatography | |
RUO | |
8522 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Flow Cytometry, Western Blot | |
OTI9H6 | |
Unconjugated | |
GAS7 | |
AW124766; B230343A10Rik; GAS7; GAS-7; Gas7-cb; growth arrest specific 7; growth arrest-specific 7; growth arrest-specific protein 7; KIAA0394; MLL/GAS7; MLL/GAS7 fusion protein | |
Full length human recombinant protein of human GAS7 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction