Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAS7 Mouse anti-Human, Clone: OTI8H10, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA501907

Description
Growth arrest-specific 7 is expressed primarily in terminally differentiated brain cells and predomitly in mature cerebellar Purkinje neurons. GAS7 plays a putative role in neuronal development. Several transcript variants encoding proteins which vary in the N-terminus have been described.Specifications
GAS7 | |
Monoclonal | |
0.7 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O60861 | |
GAS7 | |
Full length human recombit protein of human GAS7 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI8H10 | |
Unconjugated | |
GAS7 | |
growth arrest specific 7 | |
Mouse | |
Affinity Chromatography | |
RUO | |
8522 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction