Learn More
GAS7 Mouse anti-Human, Clone: OTI9H6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501873

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Growth arrest-specific 7 is expressed primarily in terminally differentiated brain cells and predomitly in mature cerebellar Purkinje neurons. GAS7 plays a putative role in neuronal development. Several transcript variants encoding proteins which vary in the N-terminus have been described.Specifications
GAS7 | |
Monoclonal | |
Unconjugated | |
GAS7 | |
growth arrest specific 7 | |
Mouse | |
Affinity Chromatography | |
RUO | |
8522 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI9H6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O60861 | |
GAS7 | |
Full length human recombit protein of human GAS7 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.