Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAS7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584663
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584663 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584663 Supplier Invitrogen™ Supplier No. PA584663
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SAKLHSEVEK PLMNFRENFK KDMKKCDHHI ADLRKQLASR YASVEKARKA LTERQRDLEM KTQQLEIKLS NKTEEDIKKG RRKS Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.

Growth arrest-specific 7 is expressed primarily in terminally differentiated brain cells and predominantly in mature cerebellar Purkinje neurons. GAS7 plays a putative role in neuronal development. Several transcript variants encoding proteins which vary in the N-terminus have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAS7
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GAS7
Gene Accession No. O60861
Gene Alias AW124766; B230343A10Rik; GAS7; GAS-7; Gas7-cb; growth arrest specific 7; growth arrest-specific 7; growth arrest-specific protein 7; KIAA0394; MLL/GAS7; MLL/GAS7 fusion protein
Gene Symbols GAS7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GAS7. Recombinant protein control fragment (Product #RP-104259).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8522
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.