Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAS8 Antibody (OTI10G2), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270795IR
Description
GAS8 Monoclonal specifically detects GAS8 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
GAS8 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GAS8 | |
Full length recombinant protein of human GAS8 (NP_001472) produced in E.coli. | |
0.1 mL | |
2622.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI10G2 | |
Western Blot | |
GAS-11, GAS11MGC138326, GAS-8, growth arrest specific 11, growth arrest-specific 11, growth arrest-specific 8, Growth arrest-specific protein 11, growth arrest-specific protein 8 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction