Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAS8 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986970
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6970 Supplier Bioss Supplier No. BS6009R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene includes 11 exons spanning 25 kb and maps to a region of chromosome 16 that is sometimes deleted in breast and prostrate cancer. The second intron contains an apparently intronless gene, C16orf3, that is transcribed in the opposite orientation. This gene is a putative tumor suppressor gene.

Specifications

Antigen GAS8
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GAS8
Gene Accession No. O95995
Gene Alias beta growth arrest specific protein 11; BOS_17043; CILD33; DRC4; Dynein regulatory complex subunit 4; GAS11; GAS-11; GAS8; GAS-8; growth arrest specific 11; growth arrest specific 8; growth arrest specific protein 11; growth arrest-specific 8; Growth arrest-specific protein 11; Growth arrest-specific protein 8; MGC138326
Gene Symbols GAS8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GAS8.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2622
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.