Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GASP1 Monoclonal Antibody (288518)
GREENER_CHOICE

Catalog No. PIMA523990
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523990 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523990 Supplier Invitrogen™ Supplier No. MA523990
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In direct ELISAs and Western blots, no cross-reactivity with recombinant human GASP-2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Growth and differentiation factor-associated serum protein-1 (GASP-1) is a secreted inhibitory TGF-beta binding protein that contains multiple protease inhibitor structural domains. It is expressed primarily in the ovary, testis, and brain, and can act as a potent soluble inhibitor of myostatin and GDF-11, but not Activin A. The GASP-1 gene encodes a 571 amino acid protein that contains a 29 amino acid secretion signal sequence, and multiple identifiable structural features, including a WAP domain, a follistatin/Kazal domain, an immunoglobulin domain, two tandem Kunitz domains, and a netrin domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GASP1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 288518
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene WFIKKN2
Gene Accession No. Q8TEU8
Gene Alias 2610304F08Rik; AY100450; Gasp1; GASP-1; Growth and differentiation factor-associated serum protein 1; hGASP-1; mGASP-1; multivalent protease inhibitor protein; RGD1305361; UNQ9235/PRO31996; WAP four-disulfide core domain 20B; WAP, follistatin, immunoglobulin, kunitz and NTR domain-containing-related protein; WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2; WAP, FS, Ig, two KU and NTR module related protein; WAP, Kazal, immunoglobulin, Kunitz and NTR domain-containing protein 2; WAP, kazal, immunoglobulin, kunitz and NTR domain-containing protein 2; WAP, Kazal, immunoglobulin, Kunitz and NTR domain-containing protein 2; WFDC20B; WFIKKN2; WFIKKN-related protein; WFIKKNRP
Gene Symbols WFIKKN2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human GASP-1/WFIKKNRP Leu35-His576.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124857
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.