Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GASP1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572067
Description
Protease-inhibitor that contains multiple distinct protease inhibitor domains. Probably has serine protease- and metalloprotease-inhibitor activity. Inhibits the biological activity of mature myostatin, but not activin (By similarity).
Specifications
GASP1 | |
Polyclonal | |
Unconjugated | |
WFIKKN2 | |
WFIKKN2; WFIKKNRP; GASP1; GASP-1; Growth and differentiation factor-associated serum protein 1; hGASP-1; WAP; follistatin; immunoglobulin; kunitz and NTR domain-containing-relatedprotein; WAP; follistatin/kazal; immunoglobulin; kunitz and netrin domain containing 2; WAP; FS; Ig; two KU and NTR module related protein; WAP; kazal; immunoglobulin; kunitz and NTR domain-containing protein 2; WFIKKN-related protein; WFIKKNRPmultivalent protease inhibitor protein | |
Rabbit | |
Protein A | |
RUO | |
124857, 278507 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.95 mg/mL | |
PBS with 0.09% sodium azide | |
Q7TQN3, Q8TEU8 | |
WFIKKN2 | |
KLH conjugated synthetic peptide between 535-564 amino acids from the C-terminal region of human WFIKKN2 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction