Learn More
Invitrogen™ GASP2 Polyclonal Antibody

Description
Immunogen sequence: STCERECSRD QDCAAAEKCC INVCGLHSCV AARFPGSPAA PTTAASCEGF VCPQQGSDCD I Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 82%.
Specifications
Specifications
| Antigen | GASP2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | WFIKKN1 |
| Gene Accession No. | Q96NZ8 |
| Gene Alias | C16orf12; GASP2; GASP-2; Growth and differentiation factor-associated serum protein 2; hGASP-2; PSEC0040; RJD2; WAP four-disulfide core domain 20A; WAP, follistatin, immunoglobulin, kunitz and NTR domain-containing protein; WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 1; WAP, FS, Ig, KU, and NTR-containing protein; WAP, Kazal, immunoglobulin, Kunitz and NTR domain-containing protein 1; WFDC20A; WFIKKN; WFIKKN1 |
| Gene Symbols | WFIKKN1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.