Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gastric Lipase Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11060930AF647
Description
Gastric Lipase Polyclonal specifically detects Gastric Lipase in Human, Mouse samples. It is validated for Western Blot.Specifications
Gastric Lipase | |
Polyclonal | |
Western Blot | |
P07098 | |
LIPF | |
A synthetic peptide within residues 1-100 of human Gastric Lipase. [Swiss-Prot# P07098] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EC 3.1.1, EC 3.1.1.3, Gastric lipase, gastric triacylglycerol lipase, GL, HGL, HLAL, lipase, gastric, MGC138477, MGC142271 | |
Rabbit | |
45 kDa | |
0.1 mL | |
Autophagy, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Endothelial Cell Markers, Glia Markers, Growth and Development, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers, Signal Transduction | |
8513 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction