Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gastric Lipase Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11060930R
Description
Gastric Lipase Polyclonal specifically detects Gastric Lipase in Human, Mouse samples. It is validated for Western Blot.Specifications
Gastric Lipase | |
Polyclonal | |
Western Blot | |
P07098 | |
LIPF | |
A synthetic peptide within residues 1-100 of human Gastric Lipase. [Swiss-Prot# P07098] | |
Affinity Purified | |
RUO | |
Primary | |
Human and mouse. Immunogen sequence has 87% homology to canine. | |
Store at 4C in the dark. |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EC 3.1.1, EC 3.1.1.3, Gastric lipase, gastric triacylglycerol lipase, GL, HGL, HLAL, lipase, gastric, MGC138477, MGC142271 | |
Rabbit | |
45 kDa | |
0.1 mL | |
Autophagy, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Endothelial Cell Markers, Glia Markers, Growth and Development, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers, Signal Transduction | |
8513 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction