Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gastrin Antibody (GAST/2635), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308703B
Description
Gastrin Monoclonal antibody specifically detects Gastrin in Human samples. It is validated for ELISASpecifications
Gastrin | |
Monoclonal | |
Biotin | |
GAS, gastrin | |
Recombinant full-length human Gastrin protein (Uniprot: P01350) | |
100 μg | |
Cancer, Cardiovascular Biology, Cell Biology, Immunology, Signal Transduction | |
2520 | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA | |
GAST/2635 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction