Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gastrin Mouse anti-Human, Clone: GAST/2633, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen Gastrin
Clone GAST/2633
Concentration 0.2 mg/ml
Dilution ELISA 1-5 ug/ml
Applications ELISA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123104
SDP
View Documents
Novus Biologicals
NBP30781520UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

Gastrin Monoclonal specifically detects Gastrin in Human samples. It is validated for ELISA.
Specifications

Specifications

Gastrin
0.2 mg/ml
ELISA
Unconjugated
Mouse
Human
GAS, gastrin
Recombinant full-length human Gastrin protein
Primary
Store at 4°C.
GAST/2633
ELISA 1-5 ug/ml
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
2520
IgG2c κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.