Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gastrin Mouse anti-Human, Clone: GAST/2634, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Gastrin |
---|---|
Clone | GAST/2634 |
Concentration | 0.2 mg/ml |
Dilution | ELISA 1-5 ug/ml, Immunohistochemistry-Paraffin |
Applications | ELISA, Immunohistochemistry (Paraffin) |
Description
Gastrin Monoclonal specifically detects Gastrin in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.Specifications
Gastrin | |
0.2 mg/ml | |
ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
GAS, gastrin | |
Recombinant full-length human Gastrin protein | |
Primary | |
Store at 4°C. |
GAST/2634 | |
ELISA 1-5 ug/ml, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
2520 | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title