Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gastrin-releasing Peptide/Bombesin/Neuromedin C Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30328220UL
Description
Gastrin-releasing Peptide/Bombesin/Neuromedin C Polyclonal specifically detects Gastrin-releasing Peptide/Bombesin/Neuromedin C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Gastrin-releasing Peptide/Bombesin/Neuromedin C | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
Unconjugated | |
BN, bombesin, gastrin-releasing peptide, GRP-10, neuromedin C, pre-progastrin releasing peptide, preproGRP, proGRP | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-148 of human Gastrin-releasing Peptide/Bombesin/Neuromedin C (NP_002082.2). | |
20 μL | |
GPCR, Neuroscience | |
2922 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction