Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gastrin-releasing Peptide R/GRPR Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP130937

Catalog No. NBP130937


Description

Description

Gastrin-releasing Peptide R/GRPR Polyclonal specifically detects Gastrin-releasing Peptide R/GRPR in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

Gastrin-releasing Peptide R/GRPR
Polyclonal
Unconjugated
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
GRPR
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human GRPR. The exact sequence is proprietary.
Affinity Purified
RUO
Primary
Not yet tested in any other species.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
1.0mg/mL
Western Blot 1:500-1:3000
gastrin-releasing peptide receptor, GRP-preferring bombesin receptor, GRP-R
Rabbit
43 kDa
100 μL
GPCR
2925
Human, Mouse, Rat
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only